Florisil® for chromatography
Supplier: Roth Carl
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
0101.1 0101.2
ROTH0101.1EA 0 EUR
ROTH0101.1 ROTH0101.2
Florisil® for chromatography
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1