Florisil® for pesticide residue analysis
Supplier: Roth Carl
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
T201.1 T201.2
ROTHT201.1EA 0 EUR
ROTHT201.1 ROTHT201.2
Florisil® for pesticide residue analysis
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1