Sodium aluminate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium Aluminium oxide, Aluminum sodium oxide, Aluminium sodium dioxide, Sodium aluminum oxide
M-Clarity™ quality level = MQ100
13404-5KG-R 13404-50KG-R 13404-2.5KG-R
SIAL13404-5KG-REA 172 EUR
SIAL13404-5KG-R SIAL13404-50KG-R SIAL13404-2.5KG-R
Sodium aluminate, Sigma-Aldrich®
Sodium aluminate
Formula: NaAlO₂
MW: 81.97 g/mol
Melting Pt: 1800 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00801010
CAS Number: 1302-42-7
EINECS: 215-100-1
UN: 2812
ADR: 8,
Merck Index: 12,08715