Human Tagged ORF Clone Lentiviral Particle, TNFRSF4
Supplier: OriGene
ORIGRC211253L3VEA 1320 EUR
ORIGRC211253L3V
Human Tagged ORF Clone Lentiviral Particle, TNFRSF4
Vectors, Plasmids and Libraries
Lenti ORF particles, TNFRSF4 (Myc-DDK tagged) - Human tumor necrosis factor receptor superfamily, member 4 (TNFRSF4), 200 µl, >10^7 TU/ml.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence. This clone is substantially in agreement with the reference, but a complete review of all prevailing variants is recommended prior to use.
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.