Tork Hand Sanitising Alcohol Foam
Supplier: Essity
Tork
SCAA520102EA 279 EUR
SCAA520102
Tork Hand Sanitising Alcohol Foam
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Dermatologically-tested, Tork hand sanitising alcohol foam is gentle on the skin.
  • Efficacy: Bacteria, yeast and fungi, mycobacteria, virus
  • Perfume free; Dye free
  • Efficacy norms: EN 1500, EN 13727, EN 13624, EN 1650, EN 14348 (tubercolosis), EN 14476 (all enveloped viruses)
Effective against a variety of problem organisms - conforms with EN 1500, EN 12791 and EN 14476 (all enveloped viruses, such as: HIV, SARS, Corona, HBV, HCV, influenza H1N1, H5N1 and more.