(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥95% (by HPLC), Sigma-Aldrich®
Supplier: Merck
Synonyms: (S,R,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine, (+)-N,N-Bis[(1R)-1-phenylethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine, (11bS)
M-Clarity™ quality level = MQ100
665363-100MG 665363-2G 665363-500MG
SIAL665363-100MGEA 60.1 EUR
SIAL665363-100MG SIAL665363-2G SIAL665363-500MG
(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥95% (by HPLC), Sigma-Aldrich®
(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine
DSM MonoPhos Family of Highly Efficient Privileged Ligands
Formula: C₃₆H₃₀NO₂P
MW: 539.60 g/mol
Melting Pt: 102-103 °C
MDL Number: MFCD08561138
CAS Number: 497883-22-4