tert-Butyl azetidine-1-carboxylate
Supplier: Thermo Fisher Scientific
Warning
467060010 467060050
467060010.EA 96.9 EUR
467060010. 467060050.
tert-Butyl azetidine-1-carboxylate
tert-Butyl azetidine-1-carboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264397
CAS Number: 147621-21-4
Order Now
Specification Test Results
Appearance clear light yellow liquid
Infrared spectrum Conforms
GC ≥96.0 %