Sodium phosphonoformate hexahydrate 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Sodium phosphonatoformate hexahydrate, Foscarnet trisodium hexahydrate
467270010 467270050
467270010.EA 219 EUR
467270010. 467270050.
Sodium phosphonoformate hexahydrate 98%
Sodium phosphonoformate hexahydrate
Formula: Na₂O₃PCO₂Na·6H₂O
MW: 300.08 g/mol (191.98 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150176
CAS Number: 34156-56-4
Order Now
Specification Test Results
Appearance white to almost white crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %