Tetrabenazine 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Isobutyl-9,10-dimethoxy-1,3,4,6,7,11b-hexahydro-2H-benzo[a]quinolizin-2-one
469990010 469992500
469990010.EA 258 EUR
469990010. 469992500.
Tetrabenazine 98%
Tetrabenazine
Formula: C₁₉H₂₇NO₃
MW: 317.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08461052
CAS Number: 58-46-8
EINECS: 200-383-6
Order Now
Specification Test Results
Appearance white to light yellow to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 124°C to 130°C