Isothiazole-5-carboxylic acid 97%
Supplier: Thermo Scientific
Warning
470360010 470362500
470360010.EA 341 EUR
470360010. 470362500.
Isothiazole-5-carboxylic acid 97%
Isothiazole-5-carboxylic acid
Formula: C₄H₃NO₂S
MW: 129.137 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07369964
CAS Number: 10271-85-9
Order Now
Specification Test Results
Appearance white to beige solid
Infrared spectrum Conforms
HPLC ≥96 %