Lornoxicam 98%
Supplier: Thermo Scientific
Danger
Synonyms: 6-Chloro-4-hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-Dioxide, 6-Chloro-4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide
472050010 472050050
472050010.EA 68.6 EUR
472050010. 472050050.
Lornoxicam 98%
Lornoxicam
Formula: C₁₃H₁₀ClN₃O₄S₂
MW: 371.81 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866163
CAS Number: 70374-39-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,05603
Order Now
Specification Test Results
Appearance white to yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %