6-Benzyladenine hydrochloride 98%
Supplier: Thermo Scientific
Warning
Synonyms: 6-Benzylaminopurine hydrochloride, N-Benzyl-9H-purin-6-amine hydrochloride
472110010 472110050 472112500
472110010.EA 180 EUR
472110010. 472110050. 472112500.
6-Benzyladenine hydrochloride 98%
6-Benzyladenine hydrochloride
Formula: C₁₂H₁₂ClN₅
MW: 261.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265275
CAS Number: 162714-86-5
Order Now
Specification Test Results
Appearance white almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %