2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, Octyl 2-Cyano-3,3-diphenylacrylate, 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octocrylene
474550250 474551000
474550250.EA 39.8 EUR
474550250. 474551000.
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98%
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate
Formula: C₂₄H₂₇NO₂
MW: 361.49 g/mol
Flash Pt: 230°C
Storage Temperature: Refrigerator
MDL Number: MFCD00059260
CAS Number: 6197-30-4
EINECS: 228-250-8
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance clear colorless to yellow liquid
Infrared spectrum Conforms
HPLC ≥97.5 %