Laboratory Washer-Disinfector, PG 8582
Supplier: Miele
MIEL9960770EA 0 EUR
MIEL9960770
Laboratory Washer-Disinfector, PG 8582
Glassware Washing Machines
Washer-disinfector with liquid dispensing and EcoDry function.
  • Performance/cycle, e.g. 8 DIN mesh trays or 42 GYN specula
  • Safety through monitoring - wash pressure and spray arm monitoring
  • Convenient drying support - EcoDry
  • Total security - link to process documentation possible