Iodomethane[¹³C] (99% ¹³C ) stabilised, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl iodide-(13C)
Stabilization: Stabilised with copper
M-Clarity™ quality level = MQ200
277185-1G 277185-5G
SIAL277185-1GEA 187 EUR
SIAL277185-1G SIAL277185-5G
Iodomethane[¹³C] (99% ¹³C ) stabilised, Sigma-Aldrich®
Iodomethane-[13C]
Formula: H₃CI
MW: 142.93 g/mol
MDL Number: MFCD00001075
CAS Number: 4227-95-6
UN: 2644
ADR: 6.1,I