IPTG (isopropyl-β-D-thiogalactopyranoside), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
I6758-10G I6758-1G I6758-5G
SIALI6758-10GEA 815 EUR
SIALI6758-10G SIALI6758-1G SIALI6758-5G
IPTG (isopropyl-β-D-thiogalactopyranoside), Sigma-Aldrich®
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0