IPTG (isopropyl-β-D-thiogalactopyranoside)
Supplier: Molekula
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
21689530-25G 21689530-5G
MOLE21689530-25GEA 545 EUR
MOLE21689530-25G MOLE21689530-5G
IPTG (isopropyl-β-D-thiogalactopyranoside)
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0