2-Bromo-3,5-dimethoxytoluene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

677736-1G 677736-5G
SIAL677736-1GEA 86 EUR
SIAL677736-1G SIAL677736-5G
2-Bromo-3,5-dimethoxytoluene, Sigma-Aldrich®
2-Bromo-3,5-dimethoxytoluene
Formula: C₉H₁₁BrO₂
MW: 231.09 g/mol
CAS Number: 13321-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR