Guaiacol

Supplier: LGC Standards PROMOCHEM

PROOMM0354.04EA 1190 EUR
PROOMM0354.04
Guaiacol
Guaiacol
Formula: C₇H₈O₂
MW: 124,14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1,1534 g/cm³ (0 °C)
Flash Pt: 82 °C
MDL Number: MFCD00002185
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR