1-(2-Thenoyl)-3,3,3-trifluoroacetone, Sigma-Aldrich®
Supplier: Merck
Synonyms: (TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone
M-Clarity™ quality level = MQ200
T27006-25G T27006-100G
SIALT27006-25GEA 70.6 EUR
SIALT27006-25G SIALT27006-100G
1-(2-Thenoyl)-3,3,3-trifluoroacetone, Sigma-Aldrich®
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formula: C₈H₅F₃O₂S
MW: 222.19 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 40…47 °C
Flash Pt: 111 °C
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7