2-Methyl-9,10-anthraquinone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methylanthraquinone, 2-Methylanthracene-9,10-Dione

M-Clarity™ quality level = MQ100

65800-100G 65800-500G
SIAL65800-100GEA 67 EUR
SIAL65800-100G SIAL65800-500G
2-Methyl-9,10-anthraquinone, Sigma-Aldrich®
2-Methyl-9,10-anthraquinone
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 236…238 °C (10 mmHg)
Melting Pt: 171…173 °C
Density: 1.365 g/cm³ (20 °C)
MDL Number: MFCD00001235
CAS Number: 84-54-8
EINECS: 201-539-6
UN: 3077
ADR: 9 III
Merck Index: 12,06100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR