Pyridoxine hydrochloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P6280-100G P6280-10G
SIALP6280-100GEA 414 EUR
SIALP6280-100G SIALP6280-10G
Pyridoxine hydrochloride, Sigma-Aldrich®
Pyridoxine hydrochloride
Formula: C₈H₁₂ClNO₃
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072