N,N-Diethyl-m-toluamide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Diethyl-m-toluamide, Diethyltoluamide, N,N-Diethyl-3-methylbenzamide, Deet
M-Clarity™ quality level = MQ100
D100951-100G
D100951-2KG
D100951-500G
SIALD100951-100GEA
50.5
EUR
SIALD100951-100G
SIALD100951-2KG
SIALD100951-500G
N,N-Diethyl-m-toluamide, Sigma-Aldrich®
N,N-Diethyl-m-toluamide
Formula:
C₁₂H₁₇NO MW: 191.27 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: –45 °C Density: 0.996 g/cm³ (20 °C) Flash Pt: 140 °C |
MDL Number:
MFCD00009046 CAS Number: 134-62-3 EINECS: 205-149-7 Merck Index: 13,02876 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...