Tetrahydrothiophen-3-one, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dihydrothiophen-3(2H)-one, 4,5-Dihydrothiophen-3(2H)-one, 4,5-Dihydro-3(2H)-thiophenone
M-Clarity™ quality level = MQ300
W326607-1KG-K
W326607-250G-K
W326607-25G-K
SIALW326607-1KG-KEA
2700
EUR
SIALW326607-1KG-K
SIALW326607-250G-K
SIALW326607-25G-K
Tetrahydrothiophen-3-one, Sigma-Aldrich®
Tetrahydrothiophen-3-one
Formula:
C₄H₆OS MW: 102.16 g/mol |
MDL Number:
MFCD00005412 CAS Number: 1003-04-9 EINECS: 213-698-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...