Tetrahydrothiophen-3-one, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dihydrothiophen-3(2H)-one, 4,5-Dihydrothiophen-3(2H)-one, 4,5-Dihydro-3(2H)-thiophenone

M-Clarity™ quality level = MQ100

264784-25G 264784-5G
SIAL264784-25GEA 31.1 EUR
SIAL264784-25G SIAL264784-5G
Tetrahydrothiophen-3-one, Sigma-Aldrich®
Tetrahydrothiophen-3-one
Formula: C₄H₆OS
MW: 102.16 g/mol
MDL Number: MFCD00005412
CAS Number: 1003-04-9
EINECS: 213-698-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR