5-Chloro-2-thiophenesulphonyl chloride

Supplier: Thermo Scientific

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

JW00059.10 JW00059.50
MAYBJW00059.10EA 0 EUR
MAYBJW00059.10 MAYBJW00059.50
5-Chloro-2-thiophenesulphonyl chloride
5-Chloro-2-thiophenesulphonyl chloride
Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 64…66 °C (0.05 mmHg)
Melting Pt: 28 °C
Density: 1.623 g/cm³ (20 °C)
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR