1,4-Bis(5-phenyloxazol-2-yl)benzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: POPOP, 1,4-Di(5-Phenyl-2-oxazolyl)benzene
M-Clarity™ quality level = MQ200
P3754-100G P3754-10G P3754-25G P3754-5G
SIALP3754-100GEA 1320 EUR
SIALP3754-100G SIALP3754-10G SIALP3754-25G SIALP3754-5G
1,4-Bis(5-phenyloxazol-2-yl)benzene, Sigma-Aldrich®
1,4-Bis(5-phenyloxazol-2-yl)benzene
Formula: C₂₄H₁₆N₂O₂
MW: 364.4 g/mol
Melting Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00005309
CAS Number: 1806-34-4
EINECS: 217-304-6