Diethoxy(dimethyl)silane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethyldiethoxysilane
M-Clarity™ quality level = MQ100
175595-500ML 175595-100ML
SIAL175595-500MLEA 212 EUR
SIAL175595-500ML SIAL175595-100ML
Diethoxy(dimethyl)silane, Sigma-Aldrich®
Diethoxy(dimethyl)silane
Formula: C₆H₁₆O₂Si
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –87 °C
Density: 0.853 g/cm³ (20 °C)
MDL Number: MFCD00009068
CAS Number: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3,II