Phenol liquified ≥75%

Supplier: PanReac AppliChem

A1624.0500EA 123 EUR
A1624.0500
Phenol liquified ≥75%
Phenol liquified
Formula: C₆H₅OH
MW: 94,11 g/mol
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 2821
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR