Oxalic acid 0,5% in sulphuric acid 50% Reag. Ph. Eur. 1061401
Supplier: VWR Chemicals
87883.290EA
256
EUR
87883.290
Oxalic acid 0,5% in sulphuric acid 50% Reag. Ph. Eur. 1061401
Oxalic acid
Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula:
C₂H₂O₄ MW: 90,04 g/mol |
MDL Number:
MFCD00002573 UN: 2796 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...