Bottles, wide neck, with standard ground stoppers
Supplier: VITLAB
100494 101594 101694 101794 101894
215-6855EA 77.8 EUR
215-6855 215-6847 215-6849 215-6850 215-6852
Bottles, wide neck, with standard ground stoppers
Bottles Plastic Bottles
PP, natural colour with PP stoppers.
  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Interchangeable ground joint stoppers
Perfect for sample collection, storage and transport. Ideal for liquids, pastes and granular material. The wide neck permits easy filling. Can be filled with hot substances.