4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Cyano-5-fluorobenzotrifluoride
B20617.06 B20617.14
B20617.06EA 84.8 EUR
B20617.06 B20617.14
4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Purity: 98+% CAS No.: 194853-86-6 Molecular Formula: C8H3F4N Formula Weight: 189.11 Melting Point: 43-45° Boiling Point: 194-196° UN No.: 3439 MDL No.: MFCD00061283
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 194…196 °C
Melting Pt: 43…45 °C
Density: 1.350
Storage Temperature: Ambient
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1,III