2,6-Dimethyl-4-heptanol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diisobutylcarbinol

M-Clarity™ quality level = MQ100

541486-1L 541486-250ML
SIAL541486-1LEA 217 EUR
SIAL541486-1L SIAL541486-250ML
2,6-Dimethyl-4-heptanol, Sigma-Aldrich®
2,6-Dimethyl-4-heptanol
Formula: C₉H₂₀O
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.8237 g/cm³ (0 °C)
MDL Number: MFCD00008944
CAS Number: 108-82-7
EINECS: 203-619-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR