Triethyl orthoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,1,1-Triethoxyethane
M-Clarity™ quality level = MQ200
75580-100ML
75580-500ML
SIAL75580-100MLEA
44.9
EUR
SIAL75580-100ML
SIAL75580-500ML
Triethyl orthoacetate, Sigma-Aldrich®
Triethyl orthoacetate
Formula:
C₈H₁₈O₃ MW: 162.23 g/mol Boiling Pt: 142 °C (1013 hPa) Density: 0.8847 g/cm³ (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00009223 CAS Number: 78-39-7 EINECS: 201-112-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...