3,5-Dichloro-4-iodobenzotrifluoride
Supplier: Thermo Scientific
SEW01636DA SEW01636EA
MAYBSEW01636DAEA 0 EUR
MAYBSEW01636DA MAYBSEW01636EA
3,5-Dichloro-4-iodobenzotrifluoride
3,5-Dichloro-4-iodobenzotrifluoride
Formula: C₇H₂Cl₂F₃I
MW: 340.9 g/mol
MDL Number: MFCD00084997
CAS Number: 175205-56-8