Anthralin, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one

M-Clarity™ quality level = MQ200

D2773-1G D2773-5G
SIALD2773-1GEA 147 EUR
SIALD2773-1G SIALD2773-5G
Anthralin, Sigma-Aldrich®
Anthralin
Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 193.5…195 °C
Density: 1.433 g/cm³ (20 °C)
MDL Number: MFCD00053409
CAS Number: 1143-38-0
EINECS: 214-538-0
Merck Index: 12,00723

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR