6-Propyl-2-thiouracil, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
P3755-100G P3755-10G P3755-25G
SIALP3755-100GEA 296 EUR
SIALP3755-100G SIALP3755-10G SIALP3755-25G
6-Propyl-2-thiouracil, Sigma-Aldrich®
6-Propyl-2-thiouracil
Formula: C₇H₁₀N₂OS
MW: 170.24 g/mol
Boiling Pt: 448
Melting Pt: 215…218 °C
MDL Number: MFCD00006041
CAS Number: 51-52-5
EINECS: 200-103-2
Merck Index: 12,08054