9-Acetylanthracene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-(9-Anthracenyl)acetone, 9-Anthryl methyl ketone, 1-(9-Anthracenyl)ethanone
M-Clarity™ quality level = MQ200
A12607-25G
A12607-5G
SIALA12607-25GEA
282
EUR
SIALA12607-25G
SIALA12607-5G
9-Acetylanthracene, Sigma-Aldrich®
9-Acetylanthracene
Formula:
C₁₆H₁₂O MW: 220.27 g/mol Boiling Pt: 374 °C (1013 hPa) Melting Pt: 82 °C Density: 1.26 g/cm³ (20 °C) |
MDL Number:
MFCD00001259 CAS Number: 784-04-3 EINECS: 212-315-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...