1,2,4-Triazole 99.5%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole
139280050 139280250 139281000 139285000
ACRO139280050EA 19.2 EUR
ACRO139280050 ACRO139280250 ACRO139281000 ACRO139285000
1,2,4-Triazole 99.5%
1,2,4-Triazole
Formula: C₂H₃N₃
MW: 69.07 g/mol
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: 119…122 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00005228
CAS Number: 288-88-0
EINECS: 206-022-9
Merck Index: 12,09735
Order Now
Specification Test Results
Appearance White Powder and/or flakes
Infrared spectrum Conforms
Melting point 119 to 122 °C
Water ≤0.2 % (K.F.)
HPLC ≥99.4 %