4-tert-Octylphenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
290823-1KG 290823-25G
SIAL290823-1KGEA 196 EUR
SIAL290823-1KG SIAL290823-25G
4-tert-Octylphenol, Sigma-Aldrich®
4-tert-Octylphenol
Formula: C₁₄H₂₂O
Boiling Pt: 279…281 °C (1013 hPa)
Melting Pt: 80…81 °C
Density: 0.932 g/cm³ (20 °C)
Flash Pt: 147 °C
MDL Number: MFCD00002368
CAS Number: 140-66-9
EINECS: 205-426-2
UN: 2430
ADR: 8,III