8-Chloroquinaldine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 8-Chloro-2-methylquinoline

M-Clarity™ quality level = MQ100

683914-1G 683914-5G
SIAL683914-1GEA 52.4 EUR
SIAL683914-1G SIAL683914-5G
8-Chloroquinaldine, Sigma-Aldrich®
8-Chloroquinaldine
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00051764
CAS Number: 3033-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR