4-Amino-2-methylquinoline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Aminoquinaldine, 2-Methylquinolin-4-amine
M-Clarity™ quality level = MQ100
A79000-25G
A79000-5G
SIALA79000-25GEA
286
EUR
SIALA79000-25G
SIALA79000-5G
4-Amino-2-methylquinoline, Sigma-Aldrich®
4-Amino-2-methylquinoline
Formula:
C₁₀H₁₀N₂ MW: 158.2 g/mol Boiling Pt: 333 Melting Pt: 168 Storage Temperature: Ambient |
MDL Number:
MFCD00006759 CAS Number: 6628-04-2 EINECS: 229-604-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...