3-Amino-1H-1,2,4-triazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Amino-1,2,4-triazole, Amitrole, 3-Amino-1H-1,2,4-triazole
M-Clarity™ quality level = MQ200
A8056-100G A8056-10G A8056-25G
SIALA8056-100GEA 231 EUR
SIALA8056-100G SIALA8056-10G SIALA8056-25G
3-Amino-1H-1,2,4-triazole, Sigma-Aldrich®
3-Amino-1H-1,2,4-triazole
Formula: C₂H₄N₄
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 149…152 °C
Density: 1.138 g/cm³ (20 °C)
MDL Number: MFCD00053362
CAS Number: 61-82-5
EINECS: 200-521-5
Merck Index: 13,00489