3,5-Diamino-1H-1,2,4-triazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,5-Diamino-1,2,4-triazole, Guanazole
M-Clarity™ quality level = MQ100
D26202-100G D26202-25G
SIALD26202-100GEA 526 EUR
SIALD26202-100G SIALD26202-25G
3,5-Diamino-1H-1,2,4-triazole, Sigma-Aldrich®
3,5-Diamino-1H-1,2,4-triazole
Formula: C₂H₅N₅
MW: 99.1 g/mol
Boiling Pt: 425
Melting Pt: 205
MDL Number: MFCD00005233
CAS Number: 1455-77-2
EINECS: 215-937-2