Label removal spray, Solvent 50
Supplier: HM Industrieservice
SOLVENT 50
291-1204EA 16.8 EUR
291-1204
Label removal spray, Solvent 50
Cleaning Agents Detergents and Surface Cleansers
Label remover for hassle-free removal of paper labels.
  • After an exposure time of just two to four minutes, most labels can be removed in one go
  • Removes labels from metal, glass, cardboard and many plastics (do not use on materials that are sensitive to solvents such as plexiglass or CD cases)
  • Does not contain any CFCs