Bags, zip seal, Nalgene®
Supplier: Thermo Fisher Scientific
6255-0508 6255-0613 6255-0913 6255-0918
216-8122EA 46.1 EUR
216-8122 216-8123 216-8124 216-8125
Bags, zip seal, Nalgene®
Bags Sample Bags
Ideal for small lab items, botanical samples and other specimens. Not recommended for storing liquids. LDPE, 101 µm.
  • Strong, transparent and waterproof
  • Heavy-duty zipper seals contents