Sterilisation Pouches and Bag Rolls
Supplier: SÜDPACK MEDICA
7600138 7600056 7600136
129-1403EA 91 EUR
129-1403 129-1419 129-1433
Sterilisation Pouches and Bag Rolls
Bags Sterilisation Pouches
Polyamide-based bags for all dry heat sterilisation applications. Made of two pre-assembled plastic sides with sterilisation display.
  • Non-separable
  • Available in different versions according to size