Twist ties, SP Bel-Art
Supplier: Bel-Art Products, a part of SP
SP Bel-Art
129-1025EA 200 EUR
129-1025
Twist ties, SP Bel-Art
Ties
Strong twist tie bag closures easier to use than string or rubber bands. Made of soft, copper-tinned, annealed iron wire sealed in extruded virgin vinyl.
  • Cut the length required and twist the ends together
  • For temperature ranges from −45 to +121 °C
Ideal to bundle hard-to-handle items like rods, wires or tubes.