Bags, biohazard, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F131600005 F131600009 F131610005 F131610009 F131620005 F131620009
129-1097EA 137 EUR
129-1097 129-4920 129-1098 129-4925 129-1099 129-4921
Bags, biohazard, SP Bel-Art
Bags Waste Disposal Bags
Ideal solution for the safe disposal of used pipettes, Petri dishes, culture plates, membrane filters and more.
  • Translucent with sterilisation indicator patch
  • Can be autoclaved at a maximum of 135 °C and discarded
  • Printed with the standard biohazard warning symbol and precautionary procedures in four languages
PP, 40 µm thick, transparent.
Meets or exceeds dart-impact test requirements of 165 grams per ASTM D1709-85.
Certifikace: Meets or exceeds dart-impact test requirements of 165 grams per ASTM D1709-85.