Biohazard waste bins, Nalgene®
Supplier: Thermo Fisher Scientific
6370-0004 6370-0005 6370-0015
129-1178EA 288 EUR
129-1178 129-3607 113-2802
Biohazard waste bins, Nalgene®
Bins Waste Bins
PP.
  • Port opening in cover for convenient one handed disposal of materials. Less exposure to infectious waste
  • One step sterilisation: No need to remove autoclavable bag
  • Leakproof side and bottom construction
Autoclavable waste containers for secondary containment of biohazardous waste materials such as needles. Holds standard size autoclavable bags. Will not rust, dent or puncture. Red universal biohazard symbol.
Comply with US OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Certifikace: Comply with US OSHAMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement